Learn More
Glass Expansion Gas Fitting with Ratchet Nebulizer Connector for Agilent 7700/7900/8800/8900
SDP
Catalog No. 501907289
Encompass_Preferred
Direct Connect (DC) Nebulizer Gas Fitting Type 13. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting (70-803-1504). For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A13- and A31- DC Nebulizers. Compatible with ICP models: Agilent 5100/5110/5800/5900 ICP-OES, Agilent 7700/7800/7850/7900/8800/8900 ICP-MS, Horiba, Thermo 6000/7000 ICP-OES and X-Series/Q/RQ/TQ ICP-MS, Analytic Jena ICP-OES, PG Instruments and Advion.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7289 Supplier Glass Expansion Supplier No. 708031105
Direct Connect (DC) Nebulizer Gas Fitting Type 13. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting (70-803-1504). For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A13- and A31- DC Nebulizers. Compatible with ICP models: Agilent 5100/5110/5800/5900 ICP-OES, Agilent 7700/7800/7850/7900/8800/8900 ICP-MS, Horiba, Thermo 6000/7000 ICP-OES and X-Series/Q/RQ/TQ ICP-MS, Analytic Jena ICP-OES, PG Instruments and Advion.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001