Learn More
Glass Expansion Nebulizer Gas Line for Agilent 4500/7500
SDP
Catalog No. 501907288
Encompass_Preferred
Direct Connect (DC) Nebulizer Gas Fitting Type 12. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A12- DC Nebulizers. Compatible with Agilent 4500/7500.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7288 Supplier Glass Expansion Supplier No. 708031086
Direct Connect (DC) Nebulizer Gas Fitting Type 12. Contains Ratchet Connector Gas Line 14-28 x 500 Assembly (70-803-1505) and ICP Gas Inlet Fitting. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for A12- DC Nebulizers. Compatible with Agilent 4500/7500.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001