Learn More
Glass Expansion Nebulizer Gas Line with 2 Ratchet Fittings
SDP
Catalog No. 501907299
Encompass_Preferred
Direct Connect (DC) Nebulizer Gas Line 14-28 x 500 Assembly with 2 Ratchet Fittings. Equivalent to 70-803-1505. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for all Glass Expansion DC Nebulizers.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-7299 Supplier Glass Expansion Supplier No. 708031278
Direct Connect (DC) Nebulizer Gas Line 14-28 x 500 Assembly with 2 Ratchet Fittings. Equivalent to 70-803-1505. For argon gas connection between DC nebulizer and instrument argon gas supply fitting. Suitable for all Glass Expansion DC Nebulizers.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001