Learn More
Glass Expansion High Pressure Nebulizer Interface Kit for use with all Glass Expansion U-Series type nebulizers. High pressure sample delivery connection for common ICP applications, HPLC-ICP-MS, LC-ICP-MS, IC-ICP-MS and Single-cell.
SDP
Catalog No. 502702987
Encompass_Preferred
High Pressure Nebulizer Interface Kit for use with all Glass Expansion U-Series type nebulizers. High pressure sample delivery connection for common ICP applications, HPLC-ICP-MS, LC-ICP-MS, IC-ICP-MS and Single-cell.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2987 Supplier Glass Expansion Supplier No. FT168X
High Pressure Nebulizer Interface Kit for use with all Glass Expansion U-Series type nebulizers. High pressure sample delivery connection for common ICP applications, HPLC-ICP-MS, LC-ICP-MS, IC-ICP-MS and Single-cell.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001