Learn More
Precision Glassblowing of Colorado NEBULIZER K-TYPE 2ML/MIN
SDP
Supplier:  Precision Glassblowing of Colorado PG50082QDAC
Encompass_Preferred
K-Type Concentric Nebulizer, 0.7 lpm Ar, 30psi, 2mL/min, Quick Disconnect, Argon Connect
Catalog No. 50-420-753