Learn More
Perkin Elmer US LLC Type C3 MEINHARD Glass Nebulizer for ELAN and NexION 300/350
Catalog No. 509045507
Encompass_Preferred
MEINHARD Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.
Catalog No. 50-904-5507 Supplier Perkin Elmer US LLC Supplier No. N8102011
MEINHARD Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.