Learn More
Perkin Elmer US LLC PEEK Mira Mist Nebulizer, 0.2-2.5 mL/min
Catalog No. 509043891
Encompass_Preferred
The Mira Mist handles most liquids, most flow ranges used in ICP and is one of the most rugged and longest life nebulizers due to its outer body being strong PEEK material. It fits regular spray chambers as a direct replacement for glass concentric nebulizers. Its features include standard pressure (45 psi), low sample flow and enhanced parallel path design.
Catalog No. 50-904-3891 Supplier Perkin Elmer US LLC Supplier No. N0775330
The Mira Mist handles most liquids, most flow ranges used in ICP and is one of the most rugged and longest life nebulizers due to its outer body being strong PEEK material. It fits regular spray chambers as a direct replacement for glass concentric nebulizers. Its features include standard pressure (45 psi), low sample flow and enhanced parallel path design.