Learn More
Perkin Elmer US LLC Type A3 MEINHARD Quartz Nebulizer for ELAN and NexION 300/350
Catalog No. 509046874
Encompass_Preferred
MEINHARD™ Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.
Catalog No. 50-904-6874 Supplier Perkin Elmer US LLC Supplier No. WE024371
MEINHARD™ Nebulizers are self-aspirating and not to be used with solutions containing hydrofluoric acid. Nebulizers require an end cap (N0680504) for use with Scott Spray Chambers.