Learn More
Perkin Elmer US LLC Nebulizer Spacer
Catalog No. NC1660663
Encompass_Preferred
This spacer is used in the Standard Plastic High Sensitivity Nebulizer Complete Assembly for PinAAcle (N3160144), Organics Plastic High Sensitivity Nebulizer Complete Assembly with Tantalum Needle for PinAAcle (N3160152), Organics Plastic High Sensitivity Nebulizer Complete Assembly with Pt/Ir Needle for PinAAcle (N3160159), and the High Sensitivity Nebulizer Complete Assembly for AAnalyst 200/400/700/800 (N3150188).
Catalog No. NC1660663 Supplier Perkin Elmer US LLC Supplier No. B3150531
This spacer is used in the Standard Plastic High Sensitivity Nebulizer Complete Assembly for PinAAcle (N3160144), Organics Plastic High Sensitivity Nebulizer Complete Assembly with Tantalum Needle for PinAAcle (N3160152), Organics Plastic High Sensitivity Nebulizer Complete Assembly with Pt/Ir Needle for PinAAcle (N3160159), and the High Sensitivity Nebulizer Complete Assembly for AAnalyst 200/400/700/800 (N3150188).