Learn More
Perkin Elmer US LLC PFA-100 MicroFlow Nebulizer with Integrated Capillary
Catalog No. 509045581
Encompass_Preferred
Integrated capillary for self-aspiration, Ideal for low flow and clean applications. Chemically resistant—Ideal for strong acids, alkalis, organics Low, spike-free background for important elements such as Fe and Ca Produces a fine aerosol for high transport efficiency and high sensitivity Direct analysis of volatile and non-volatile organic solvents
Catalog No. 50-904-5581 Supplier Perkin Elmer US LLC Supplier No. N8122351
Integrated capillary for self-aspiration, Ideal for low flow and clean applications. Chemically resistant—Ideal for strong acids, alkalis, organics Low, spike-free background for important elements such as Fe and Ca Produces a fine aerosol for high transport efficiency and high sensitivity Direct analysis of volatile and non-volatile organic solvents