Learn More
New England Biolabs, Inc. NEBNext rRNA Depletion Kit (Bacteria) – 6 reactions
SDP
Catalog No. 502024028
Encompass
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic).
Catalog No. 50-202-4028 Supplier New England Biolabs, Inc. Supplier No. E7850S
The NEBNext rRNA Depletion Kit (Bacteria) employs the NEBNext RNase H-based RNA depletion workflow to target removal of rRNA from gram-positive and gram-negative organisms. The method is effective with intact and degraded RNA, from monocultures or samples with mixed bacterial species (e.g., metatranscriptomic).