Learn More
Selleck Chemical LLC Nec-1s (7-Cl-O-Nec1) 5mg 852391-15-2 Necrostatin-1 stable
SDP
Supplier:  Selleck Chemical LLC S86415MG
Encompass_Preferred
Nec-1s (7-Cl-O-Nec1, Necrostatin-1 stable) is a stable variant of Necrostatin-1. It is a more specific RIPK1 inhibitor lacking the IDO-targeting effect, with >1000-fold more selective for RIPK1 than for any other kinase out of 485 human kinases. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8109