Learn More
American Radiolabeled Chemicals Inc NECA 3H 250UC
SDP
Supplier:  American Radiolabeled Chemicals Inc ART 1773250UC
Encompass
Ethylcarboxamidoadenosine, 6'-N-adenine-2,3-3H 250uCi in ethanol:water (7:3), 1 mCi/ml, S.A. 15-30 Ci/mmol, M.W. 308.28, Shipped in Dry Ice
Catalog No. 50-901-112