Learn More
Aviva Systems Biology Corp NECAB3 Antibody - middle region (ARP57670_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57670_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against NECAB3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human NECAB3. Alternate names: APBA2BP, EFCBP3, NIP1, STIP3, SYTIP2, XB51, dJ63M2.4, dJ63M2.5ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-62926