Learn More
Aviva Systems Biology Corp Necap1 Antibody - N-terminal region (ARP55273_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55273_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Necap1. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: 1200016B17Rik, AI747569, FLJ00061, mFLJ00061ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-020