Learn More
ABclonal Technology NECAP2 Rabbit pAb
SDP
Supplier:  ABclonal Technology A143461000UL
Encompass_Preferred
This gene likely encodes a member of the adaptin-ear-binding coat-associated protein family. Studies of a similar protein in rat suggest a role in clathrin-mediated endocytosis. Alternatively spliced transcript variants have been described.
Catalog No. 50-001-06238