Learn More
Mitegen Replacement Necktube Core For IC Biomedical CX100 Dryshipper
SDP
Catalog No. NC3237528
Encompass
This replaceable necktube core for the IC Biomedical CX100 Cryo Express dry shipper. It plays a critical role in maintaining the integrity and long-term performance of your dry shipper by providing crucial insulation and ensuring optimal vacuum conditions. A fully functional necktube core is vital for minimizing liquid nitrogen evaporation, thereby preserving the cryogenic environment necessary for the safe storage and transport of sensitive biological samples. Replacing a worn or damaged necktube core can significantly extend the lifespan and efficiency of your CX100 dry shipper, preventing temperature fluctuations and sample degradation. This component ensures your dry shipper continues to meet demanding laboratory standards for cryogenic sample management. It is an indispensable part for any user relying on the CX100 for dependable cryogenic transport.
Catalog No. NC3237528 Supplier Mitegen Supplier No. TWR0059C21
This replaceable necktube core for the IC Biomedical CX100 Cryo Express dry shipper. It plays a critical role in maintaining the integrity and long-term performance of your dry shipper by providing crucial insulation and ensuring optimal vacuum conditions. A fully functional necktube core is vital for minimizing liquid nitrogen evaporation, thereby preserving the cryogenic environment necessary for the safe storage and transport of sensitive biological samples. Replacing a worn or damaged necktube core can significantly extend the lifespan and efficiency of your CX100 dry shipper, preventing temperature fluctuations and sample degradation. This component ensures your dry shipper continues to meet demanding laboratory standards for cryogenic sample management. It is an indispensable part for any user relying on the CX100 for dependable cryogenic transport.