Learn More
Med Vet International Necropsy Knife, Curved, 6", J0515, Each
Catalog No. 5000585036
Encompass_Preferred
These high-quality molybdenum-hardened stainless steel bladed knives come with a yellow nylon no-slip grip handle. German-made.
Catalog No. 50-005-85036 Supplier Med Vet International Supplier No. JOR515
These high-quality molybdenum-hardened stainless steel bladed knives come with a yellow nylon no-slip grip handle. German-made.