Learn More
Med Vet International Necropsy Knife, Narrow Blade, 5", J0507A
Catalog No. 5000585034
Encompass_Preferred
These high quality molybdenum-hardened stainless steel bladed knives come with a yellow nylon no-slip grip handle. German-made.
Catalog No. 50-005-85034 Supplier Med Vet International Supplier No. JOR507A
Add to Cart
Add to Cart
These high quality molybdenum-hardened stainless steel bladed knives come with a yellow nylon no-slip grip handle. German-made.