Learn More
Selleck Chemical LLC Necrostatin-1 10mg 4311-88-0
SDP
Supplier:  Selleck Chemical LLC S803710MG
Encompass_Preferred
Necrostatin-1 is a specific RIP1 (RIPK1) inhibitor and inhibits TNF-?-induced necroptosis with EC50 of 490 nM in 293T cells. Necrostatin-1 also blocks IDO and suppresses autophagy and apoptosis. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1092553