Learn More
Apexbio Technology LLC Necrostatin-1, 4311-88-0, MFCD00056916, 10 mg
SDP
Supplier:  Apexbio Technology LLC A421310
Encompass_Preferred
MF: C13H13N3OS, MW: 259.33, Solubility: 12.97 mg/mL in DMSO, 13.29 mg/mL in EtOH with ultrasonic,insoluble in H2O. Necroptosis is a cellular mechanism of necrotic cell death induced by apoptotic stimuli in the form of death domain receptor engagement by their respective ligands under conditions where apoptotic execution is prevented. Necrostatin-1, identified as a small-molecule inhibitor of necroptosis, is also a selective allosteric inhibitor of the death domain receptorassociated adaptor kinase RIP1.
Catalog No. 50-101-0399