Learn More
AdipoGen Necrostatin-1, 4311-88-0, MFCD00056916, 5 mg
SDP
Supplier:  AdipoGen AGCR12900M005
Encompass_Preferred
MF: C13H13N3OS, MW: 259.3, ≥98%, Appearance: Off-white to yellow solid. Competitive indoleamine 2,3-dioxygenase (IDO) inhibitor. Cell permeable, potent and selective necroptosis (a non-apoptotic form of programmed cell death) inhibitor. Showed neuroprotection in a murine model of ischemic brain injury in vivo. Selective and ATP-competitive receptor-interacting protein kinase 1 (RIPK1) inhibitor. Used in inflammatory and degenerative disease models to target RIPK1. Suppresses autophagy and apoptosis in murine brain injury models. Cardioprotective and neuroprotective.
Catalog No. 50-596-239