Learn More
Medchemexpress LLC HY-15760 50mg , Necrostatin-1 CAS:4311-88-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1576050MG
Encompass_Preferred
HY-15760 50mg Necrostatin-1 CAS: 4311-88-0 Necrostatin-1 (Nec-1) is a potent and cross the blood-brain barrier necroptosis inhibitor with an EC50 of 490 nM in Jurkat cells. Necrostatin-1 inhibits RIP1 kinase (EC50=182 nM). Necrostatin-1 is also an IDO inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9540