Learn More
TARGETMOL CHEMICALS INC NECROSTATIN-1 50MG
SDP
Catalog No. 502581078
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Necrostatin-1 (Necrostatin 1) is a specific RIP1 inhibitor and inhibits TNF-(alpha)-induced necroptosis (EC50 490 nM in Jurkat cells). purity: 99%
Catalog No. 50-258-1078 Supplier TARGETMOL CHEMICALS INC Supplier No. T184750MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Necrostatin-1 (Necrostatin 1) is a specific RIP1 inhibitor and inhibits TNF-(alpha)-induced necroptosis (EC50 490 nM in Jurkat cells). purity: 99%