Learn More
Cayman Chemical Necrostatin-1 Inactive Control, Purity: ≥98%, CAS: 64419-92-7, 1 mg
SDP
Catalog No. 502983820
Encompass_Preferred
A demethylated variant of necrostatin-1 that inhibits IDO; ~ 100-fold less effective than Nec-1 in inhibiting RIP1 kinase in vitro and 10-fold less potent than Nec-1 in a mouse necroptosis assay; inhibits TNF-induced systemic inflammatory response syndrome in vivo
Catalog No. 50-298-3820 Supplier Cayman Chemical Supplier No. 211921 MG
A demethylated variant of necrostatin-1 that inhibits IDO; ~ 100-fold less effective than Nec-1 in inhibiting RIP1 kinase in vitro and 10-fold less potent than Nec-1 in a mouse necroptosis assay; inhibits TNF-induced systemic inflammatory response syndrome in vivo