Learn More
Abcam Necrostatin-1, necroptosis inhibitor, 50MG
Catalog No. 502583374
Encompass_Preferred
MW 259.33 Da, Purity >99%. Potent, specific necroptosis inhibitor (EC₅₀ = 490 nM). Selective allosteric inhibitor of death domain receptor-associated adaptor kinase RIP1 in vitro (EC₅₀ = 182 nM). Caspase independent, programmed necrosis pathway.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3374 Supplier Abcam Supplier No. AB14105350MG
MW 259.33 Da, Purity >99%. Potent, specific necroptosis inhibitor (EC₅₀ = 490 nM). Selective allosteric inhibitor of death domain receptor-associated adaptor kinase RIP1 in vitro (EC₅₀ = 182 nM). Caspase independent, programmed necrosis pathway.
The product is subject to the following: Abcam Restricted Use Statement