Learn More
Cayman Chemical Necrostatin-1, 4311-88-0, 10 mg
SDP
Supplier:  Cayman Chemical 1165810
Encompass_Preferred
Molecular Formula C13H13N3OS, Purity ≥98%, Formula Weight 259.3, Synonyms: Nec-1; An inhibitor of RIP1 kinase that prevents the death of TNF--treated FADD-deficient Jurkat cells with an EC50 value of 490 nM.
Catalog No. NC1355361
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.