Learn More
Medchemexpress LLC HY-14622 5mg Medchemexpress, Necrostatin 2 CAS:852391-19-6 Purity:>98%
SDP
Catalog No. 501932703
Encompass_Preferred
Medchemexpress, HY-14622 5mg Necrostatin 2 CAS:852391-19-6 Necrostatin 2 is a potent necroptosis inhibitor. EC 50 for inhibition of necroptosis in FADD-deficient Jurkat T cells treated with TNF-α is 0.05 μM. Necrostatin 2 is also a RIPK1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2703 Supplier Medchemexpress LLC Supplier No. HY146225MG
Medchemexpress, HY-14622 5mg Necrostatin 2 CAS:852391-19-6 Necrostatin 2 is a potent necroptosis inhibitor. EC 50 for inhibition of necroptosis in FADD-deficient Jurkat T cells treated with TNF-α is 0.05 μM. Necrostatin 2 is also a RIPK1 inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.