Learn More
TARGETMOL CHEMICALS INC NECROSTATIN-5 5MG
SDP
Catalog No. 502580137
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Necrostatin-5 (Nec-5) is a selective inhibitor of RIP1 kinase and a cell necrosis inhibitor that prevents the death of Fadd-deficient Jurkat cells treated with TNF-(alpha) with an EC50 value of 240 nM. purity: 98%
Catalog No. 50-258-0137 Supplier TARGETMOL CHEMICALS INC Supplier No. T375585MG
Also available in 10 mg 25 mg 50 mg and bulk. Please contact Fisher for quotes. Necrostatin-5 (Nec-5) is a selective inhibitor of RIP1 kinase and a cell necrosis inhibitor that prevents the death of Fadd-deficient Jurkat cells treated with TNF-(alpha) with an EC50 value of 240 nM. purity: 98%