Learn More
AdipoGen Necrostatin-5 Nec-5
SDP
Catalog No. 502562376
Encompass_Preferred
Chemical. CAS 337349-54-9. Formula C19H17N3O2S2. MW 383.5. Necroptosis inhibitor. Indirect RIP1 kinase inhibitor. Cardioprotective
Catalog No. 50-256-2376 Supplier AdipoGen Supplier No. AGCR10049M025
Chemical. CAS 337349-54-9. Formula C19H17N3O2S2. MW 383.5. Necroptosis inhibitor. Indirect RIP1 kinase inhibitor. Cardioprotective