Learn More
AdipoGen Necrostatin-7 (25 mg)
SDP
Supplier:  AdipoGen AGCR10054M025
Encompass_Preferred
Chemical. CAS: 351062-08-3. Formula: C16H10FN5OS2. MW: 371.4. Necroptosis inhibitor. Does not inhibit RIP1 kinase.
Catalog No. 50-205-3809