Learn More
AdipoGen Necrostatin-7 Nec-7
SDP
Catalog No. 502562373
Encompass_Preferred
Chemical. CAS 351062-08-3. Formula C16H10FN5OS2. MW 371.4. Necroptosis inhibitor. Does not inhibit RIP1 kinase.
Catalog No. 50-256-2373 Supplier AdipoGen Supplier No. AGCR10054M005
Chemical. CAS 351062-08-3. Formula C16H10FN5OS2. MW 371.4. Necroptosis inhibitor. Does not inhibit RIP1 kinase.