Learn More
Enzo Life Sciences Necrostatin-7 (5mg). CAS: 351062-08-3
SDP
Supplier:  Enzo Life Sciences ALX430170M005
Encompass_Preferred
Necroptosis inhibitor. The biological activity of necrostatin-7 (Nec-7) is different from that of necrostatin-1 to -5 (Nec-1 to -5), as Nec-7 does not inhibit RIP1 kinase. Nec-7 may target an additional regulatory molecule in the pathway. Purity: ≥98% (HPLC, TLC). Appearance: Yellow solid. Solubility: Soluble in DMSO or 100% ethanol. Long Term Storage: -20°C.
Catalog No. 50-200-9259