Learn More
AdipoGen Necrosulfonamide
SDP
Supplier:  AdipoGen AGCR13705M025
Encompass_Preferred
Chemical. CAS 1360614-48-7. Formula C18H15N5O6S2. MW 461.5. Inhibitor of necroptosis, by blocking mixed lineage kinase domain-like protein MLKL, a key substrate of receptor-interacting serine-threonine kinase 3 RIP3. Shown to prevent the MLKL-RIP1-RIP3 necrosome complex from interacting with downstream necrosis effectors. Potent inhibitor of Gasdermin D that works well for mice studies. It binds directly to Gasdermin D and inhibits the oligomerization of the N-terminus and therefore the pore formation and pyroptosis. Displayed neuroprotective effects after ischemic brain injury, prevented cystine-starvation-induced necroptosis and ferroptosis in human triple negative breast cancer cells and rescued cells from TLR3 ligand-induced death.
Catalog No. 50-256-2196