Learn More
Cayman Chemical Necrosulfonamide, 1360614-48-7, 10 mg
SDP
Supplier:  Cayman Chemical 2084410
Encompass_Preferred
Inhibits necroptosis by blocking MLKL, a key substrate of RIP3
Catalog No. NC1558121