Learn More
TARGETMOL CHEMICALS INC NECROSULFONAMIDE 25MG
SDP
Catalog No. 502576351
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (E/Z)-Necrosulfonamide is a novel inhibitor of MLKL. purity: 98%
Catalog No. 50-257-6351 Supplier TARGETMOL CHEMICALS INC Supplier No. T712925MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. (E/Z)-Necrosulfonamide is a novel inhibitor of MLKL. purity: 98%