Learn More
Medchemexpress LLC Necrosulfonamide-d4 | 1795144-22-7 | >99.0% | 465.50 g/mol | C18H11D4N5O6S2 | 10 MG
SDP
Catalog No. 5000211584
Encompass_Preferred
Necrosulfonamide-d4 is a deuterium-labeled analogue of necrosulfonamide intended for use as an analytical standard, tracer, or mechanistic probe in studies of necroptosis and pyroptosis. The compound is supplied as a high-purity stable isotope (d4), with a molecular weight of approximately 465.5 g/mol and CAS number 1795144-22-7, provided in small vials suitable for analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry applications.
  • High isotopic purity (>99% deuterated forms) for reliable quantitation.
  • Compatible with LC-MS and GC-MS analytical methods.
  • Enables mechanistic studies of necroptosis and pyroptosis pathways.
  • Supplied in small-mass vials suitable for analytical use.
Catalog No. 50-002-11584 Supplier Medchemexpress LLC Supplier No. HY100573S10MG
Necrosulfonamide-d4 is a deuterium-labeled analogue of necrosulfonamide intended for use as an analytical standard, tracer, or mechanistic probe in studies of necroptosis and pyroptosis. The compound is supplied as a high-purity stable isotope (d4), with a molecular weight of approximately 465.5 g/mol and CAS number 1795144-22-7, provided in small vials suitable for analytical workflows.
  • Deuterium-labeled internal standard for mass spectrometry applications.
  • High isotopic purity (>99% deuterated forms) for reliable quantitation.
  • Compatible with LC-MS and GC-MS analytical methods.
  • Enables mechanistic studies of necroptosis and pyroptosis pathways.
  • Supplied in small-mass vials suitable for analytical use.