Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Necrosulfonamide-d4 | 1795144-22-7 | 99.3% | 465.50 g/mol | C18H11D4N5O6S2 | 1 MG
SDP

Catalog No. 5000213433
Encompass_Preferred

Necrosulfonamide-d4 is the deuterium-labeled isotopologue of necrosulfonamide supplied for research use. It is intended for use as a mass spectrometry internal standard and in biochemical or cell-based studies that probe necroptosis and pyroptosis pathways involving MLKL and Gasdermin D. The compound is provided as a characterized, high-purity research reagent for analytical and biological workflows.

  • Deuterium-labeled isotopologue suitable for mass spectrometry internal standard use.
  • Used to study MLKL- and Gasdermin D-mediated necroptosis and pyroptosis pathways.
  • High purity for reliable analytical and biochemical results.
  • Supplied in milligram-scale quantities for analytical and experimental workflows.
  • Molecular weight 465.50 g/mol; formula C18H11D4N5O6S2.

Catalog No. 50-002-13433 Supplier Medchemexpress LLC Supplier No. HY100573S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Necrosulfonamide-d4 is the deuterium-labeled isotopologue of necrosulfonamide supplied for research use. It is intended for use as a mass spectrometry internal standard and in biochemical or cell-based studies that probe necroptosis and pyroptosis pathways involving MLKL and Gasdermin D. The compound is provided as a characterized, high-purity research reagent for analytical and biological workflows.

  • Deuterium-labeled isotopologue suitable for mass spectrometry internal standard use.
  • Used to study MLKL- and Gasdermin D-mediated necroptosis and pyroptosis pathways.
  • High purity for reliable analytical and biochemical results.
  • Supplied in milligram-scale quantities for analytical and experimental workflows.
  • Molecular weight 465.50 g/mol; formula C18H11D4N5O6S2.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.