Learn More
Enzo Life Sciences Necrox-2 (1mg)
SDP
Supplier:  Enzo Life Sciences ALX430166M001
Encompass_Preferred
Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1µM NecroX™-2 prevented ~50% cell death in H9C2 cells exposed to 400µM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro. Alternative name: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(1-methanesulfonyl-piperidin-4-yl)-amine. Purity: ≥95% (NMR, LCMS). Appearance: Gray powder. Solubility: Soluble in DMSO (>10mg/ml). Long Term Storage: +4°C.
Catalog No. 50-200-9254