Learn More
Enzo Life Sciences Necrox-5. hydrochloride (1mg)
SDP
Catalog No. 502009262
Encompass_Preferred
Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1μM NecroX™-5 prevented ~50% cell death in H9C2 cells exposed to 400μM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro, as well as CCl4 -induced acute liver injury and chronic liver fibrosis in rodent models. Alternative name: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine. Purity: ≥96% (HPLC, TLC). Appearance: Yellow solid. Solubility: Soluble in DMSO (>10mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9262 Supplier Enzo Life Sciences Supplier No. ALX430247M001
Add to Cart
Add to Cart
Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1μM NecroX™-5 prevented ~50% cell death in H9C2 cells exposed to 400μM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro, as well as CCl4 -induced acute liver injury and chronic liver fibrosis in rodent models. Alternative name: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine. Purity: ≥96% (HPLC, TLC). Appearance: Yellow solid. Solubility: Soluble in DMSO (>10mg/ml). Long Term Storage: -20°C.