Learn More
Enzo Life Sciences Necrox-5 . methanesulfonate (5mg)
SDP
Supplier:  Enzo Life Sciences ALX430167M005
Encompass_Preferred
Cell permeable necrosis inhibitor with antioxidant activity. It localizes mostly in the mitochondria. Selectively blocks oxidative stress-induced necrotic cell death (0.1μM NecroX™-5 prevented ~50% cell death in H9C2 cells exposed to 400μM t-BuOOH for 2 hours LDH assay used). Does not protect against staurosporine or etoposide-induced apoptosis. Protects cells against cold shock, hypoxia and oxidative stress in vitro, as well as CCl4 -induced acute liver injury and chronic liver fibrosis in rodent models. Alternative name: [5-(1,1-Dioxo-thiomorpholin-4-ylmethyl)-2-phenyl-1H-indol-7-yl]-(tetrahydro-pyran-4-ylmethyl)-amine. Purity: ≥95% (HPLC). Solubility: Soluble in DMSO.  . Long Term Storage: -20°C.
Catalog No. 50-200-9256