Learn More
ACROBiosystems ACROBiosystems Human Nectin-1 / PVRL1 / CD111 Protein, Fc Tag
Catalog No. 501956538
Encompass_Preferred
Recombinant Protein;100UG;Human Nectin-1, Fc Tag (PV1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession # Q15223-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-3, His Tag (Cat. No. PV3-H52E4) at 5 μg/mL (100 μL/well) can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 0.6-20 ng/mL (QC tested).;Immobilized Biotinylated Human Nectin-4, His,Avitag (Cat. No. NE4-H82E7) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 2-40 ng/mL (Routinely tested).
Catalog No. 50-195-6538 Supplier ACROBiosystems Supplier No. PV1H5253100UG
Recombinant Protein;100UG;Human Nectin-1, Fc Tag (PV1-H5253) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Thr 334 (Accession # Q15223-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.4 kDa. The protein migrates as 66-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-3, His Tag (Cat. No. PV3-H52E4) at 5 μg/mL (100 μL/well) can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 0.6-20 ng/mL (QC tested).;Immobilized Biotinylated Human Nectin-4, His,Avitag (Cat. No. NE4-H82E7) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 2-40 ng/mL (Routinely tested).