Learn More
Genscript Corporation Nectin-1/PVRL1/CD111Biotin
Supplier:  Genscript Corporation Z04563500
Encompass
Nectins are a small family of Ca -independent immunoglobulin (Ig)-like cell adhesion molecules (CAMs) that organize intercellular junctions They are highly homologous to the human receptor for poliovirus and as such have been alternately named poliovirus receptor-related proteins Nectin-1 promotes cell-cell contacts by forming homophilic or heterophilic trans-dimers Heterophilic interactions have been detected between Nectin-1 and Nectin-3 and between Nectin-1 and Nectin-4 Has some neurite outgrowth-promoting activity
Catalog No. 50-000-57129