Learn More
Genscript Corporation Nectin-2/CD112Biotin His
Catalog No. 5000058271
Encompass
Nectin-2 is an adhesion molecule that has been reported to play a role in tumor growth metastasis and tumor angiogenesis Nectin-2 expression in ovarian cancer may support tumor cell adhesion leading to growth and lymph node metastasis Effect of VEGF on Nectin-2 expression as well as permeability was investigated in HUVEC
Catalog No. 50-000-58271 Supplier Genscript Corporation Supplier No. Z04564100
Nectin-2 is an adhesion molecule that has been reported to play a role in tumor growth metastasis and tumor angiogenesis Nectin-2 expression in ovarian cancer may support tumor cell adhesion leading to growth and lymph node metastasis Effect of VEGF on Nectin-2 expression as well as permeability was investigated in HUVEC