Learn More
ACROBiosystems ACROBiosystems Human Nectin-2 / CD112 Protein, His Tag
Supplier:  ACROBiosystems PV2H52E21MG
Encompass_Preferred
Recombinant Protein;1MG;Human Nectin-2, His Tag (PV2-H52E2) is expressed from human 293 cells (HEK293). It contains AA Gln 32 - Leu 360 (Accession # Q92692-2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 37.4 kDa. The protein migrates as 45-50 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PVRIG, Mouse IgG2a Fc Tag, low endotoxin (Cat. No. PVG-H5253) at 2 μg/mL (100 μL/well) can bind Human Nectin-2, His Tag (Cat. No. PV2-H52E2) with a linear range of 0.078-0.625 μg/mL (QC tested).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Human Nectin-2, His Tag (Cat. No. PV2-H52E2) with a linear range of 0.625-5 μg/mL (Routinely tested).
Catalog No. 50-195-7296