Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Nectin-3 / CD113 Protein, Fc,Avitag™
Catalog No. 501957387
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Nectin-3, Fc,Avitag (PV3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 66.1 kDa. The protein migrates as 90-95 kDa and 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-1, His Tag (Cat. No. PV1-H5223) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-3, Fc,Avitag (Cat. No. PV3-H82F3) with a linear range of 0.078-0.625 μg/mL (QC tested).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-3, Fc,Avitag (Cat. No. PV3-H82F3) with a linear range of 0.102-4 μg/mL (Routinely tested).
Catalog No. 50-195-7387 Supplier ACROBiosystems Supplier No. PV3H82F3200UG
Recombinant Protein;200UG;Biotinylated Human Nectin-3, Fc,Avitag (PV3-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3-1).;This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 66.1 kDa. The protein migrates as 90-95 kDa and 100-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-1, His Tag (Cat. No. PV1-H5223) at 2 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-3, Fc,Avitag (Cat. No. PV3-H82F3) with a linear range of 0.078-0.625 μg/mL (QC tested).;Immobilized Human TIGIT, Fc Tag (Cat. No. TIT-H5254) at 10 μg/mL (100 μL/well) can bind Biotinylated Human Nectin-3, Fc,Avitag (Cat. No. PV3-H82F3) with a linear range of 0.102-4 μg/mL (Routinely tested).