Learn More
ACROBiosystems ACROBiosystems Human Nectin-3 / CD113 Protein, His Tag
Catalog No. 501957302
Encompass_Preferred
Recombinant Protein;1MG;Human Nectin-3, His Tag (PV3-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.6 kDa. The protein migrates as 60-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-3, His Tag (Cat. No. PV3-H52E4) at 2 μg/mL (100 μL/well) can bind Human PVRL1, Fc Tag with a linear range of 2-20 ng/mL (QC tested).
Catalog No. 50-195-7302 Supplier ACROBiosystems Supplier No. PV3H52E41MG
Recombinant Protein;1MG;Human Nectin-3, His Tag (PV3-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gly 58 - Asp 400 (Accession # Q9NQS3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 39.6 kDa. The protein migrates as 60-75 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-3, His Tag (Cat. No. PV3-H52E4) at 2 μg/mL (100 μL/well) can bind Human PVRL1, Fc Tag with a linear range of 2-20 ng/mL (QC tested).