Learn More
ACROBiosystems ACROBiosystems Human Nectin-4 Protein, Fc Tag
Supplier:  ACROBiosystems NE4H52551MG
Encompass_Preferred
Recombinant Protein;1MG;Human Nectin-4, Fc Tag (NE4-H5255) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 60.5 kDa. The protein migrates as 65-70 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-4, Fc Tag (Cat. No. NE4-H5255) at 5 μg/mL (100 μL/well) can bind Human Nectin-1, His Tag (Cat. No. PV1-H5223) with a linear range of 0.313-5 μg/mL (QC tested).
Catalog No. 50-195-6409