Learn More
ACROBiosystems ACROBiosystems Human Nectin-4 Protein, His Tag
Catalog No. 501957154
Encompass_Preferred
Recombinant Protein;1MG;Human Nectin-4, His Tag (NE4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-4, His Tag (Cat. No. NE4-H52H3) at 5 μg/mL (100 μL/well) can bind Human PVRL1, Fc Tag with a linear range of 0.01-0.313 μg/mL (QC tested).
Catalog No. 50-195-7154 Supplier ACROBiosystems Supplier No. NE4H52H31MG
Recombinant Protein;1MG;Human Nectin-4, His Tag (NE4-H52H3) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 40 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human Nectin-4, His Tag (Cat. No. NE4-H52H3) at 5 μg/mL (100 μL/well) can bind Human PVRL1, Fc Tag with a linear range of 0.01-0.313 μg/mL (QC tested).