Learn More
ACROBiosystems ACROBiosystems Biotinylated Human Nectin-4 Protein, His,Avitag™
Catalog No. 501956568
Encompass_Preferred
Recombinant Protein;200UG;Biotinylated Human Nectin-4, His,Avitag (NE4-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.8 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Nectin-4, His,Avitag (Cat. No. NE4-H82E7) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 2-40 ng/mL (QC tested).
Catalog No. 50-195-6568 Supplier ACROBiosystems Supplier No. NE4H82E7200UG
Recombinant Protein;200UG;Biotinylated Human Nectin-4, His,Avitag (NE4-H82E7) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # Q96NY8-1).;This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 37.8 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Biotinylated Human Nectin-4, His,Avitag (Cat. No. NE4-H82E7) at 1 μg/mL (100 μL/well) on streptavidin precoated (0.5 μg/well) plate, can bind Human Nectin-1, Fc Tag (Cat. No. PV1-H5253) with a linear range of 2-40 ng/mL (QC tested).